Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Export
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Cy3B Tetrazine
Cy3B Tetrazine
Cy3B Tetrazine
Cy3B Tetrazine
Catalog # AATB948
Supplier:
AAT Bioquest
New Product
CAS Number:
Expand image
Cy3B Tetrazine
Catalog # AATB948
Supplier:
AAT Bioquest
New Product
CAS Number:
Specifications
Pk:
1 mg
Seq:
2070
Specifications
About this item
Product Details
:
:
729.86 g/mol
Product Details