0,00 €
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Export
ContactUsLinkComponent
Cal-520® azide
Cal-520® azide
Cal-520® azide
Catalog # AATB20613
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Expand image
Cal-520® azide
Catalog # AATB20613
Supplier:
AAT BIOQUEST
New Product
CAS Number:
Specifications
Pk:
100 µG
Seq:
1286
Specifications
About this item
Product Details
:
:
878.71 g/mol
Product Details