Order Entry
Export
ContactUsLinkComponent
Anti-NTH1 Rabbit Polyclonal Antibody
Anti-NTH1 Rabbit Polyclonal Antibody
Catalog # ANTIA10002-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-NTH1 Rabbit Polyclonal Antibody
Catalog # ANTIA10002-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Bifunctional DNA N glycoslyase/DNA (apurinic or apyrimidinic site) lyase
  • Antigen symbol:
    NTH1
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P78549
  • Antigen synonyms:
    nth endonuclease III like 1 (E. coli)|NTH endonuclease III Like 1|DNA glycoslyase/AP lyase|Endonuclease III like protein 1|Nthl1|NTHL 1|Endonuclease III-like protein 1|NTH 1|hNTH1
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,02% Sodium Azide
  • Molecular weight:
    37 kDa
  • Sequence:
    MCSPQESGMTALSARMLTRSRSLGPGAGPRGCREEPGPLRRREAAAEARKSHSPVKRPRKAQRLRVAYEGSDSEKGEGAEPLKVPVWEPQDWQQQLVNIRAMRNkDaPVDHLGTEHCYDSSAPPKVRRYQVLLSLMLSSQTKDQVTAGAMQRLRARGLTVDSILQTDDATLGKLIYPVGFWRSKVKYIKQTSAILQQHYGGDIPASVAELVALPGVGPKMAHLAMAVAWGTVSGIAVDTHVHRIANRLRWTKKATKSPEETRAALEEWLPRELWHEING
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 1 - 280 of human NTHL1 (NP_002519.1)
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to NTH1 for WB with samples derived from human.

  • Validated Applications: WB
  • Recommended Dilutions: WB: 1:500 to 1:2000

Type: Primary
Antigen: NTH1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human