Order Entry
Export
ContactUsLinkComponent
 
Anti-Collectin 12 Mouse Monoclonal Antibody [clone: 53]
  ANTIA101672-100
New Product
Anti-Collectin 12 Mouse Monoclonal Antibody [clone: 53]
  ANTIA101672-100
New Product

 

  • Antibody type:
    Primary
  • Antigen symbol:
    Collectin 12
  • Clonality:
    Monoclonal
  • Clone:
    53
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Hamster,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q5KU26
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 50% Glycerol
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant human Collectin 12 protein (a.a. 590-742), corresponding to the carbohydrate recognition domain (CRD), expressed in E. coli
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal (53) antibody to Collectin 12 for WB, IF and ELISA with samples derived from human, mouse, rat and hamster.

  • Validated Applications: WB, IF, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:500

Type: Primary
Antigen: Collectin 12
Clonality: Monoclonal
Clone: 53
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human;Mouse;Rat;Hamster