Anti-NKG2A Rabbit Polyclonal Antibody
ANTIA101003-100
New Product
- Antibody type:Primary
- Antigen name:C lectin type II protein
- Antigen symbol:NKG2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P26715
- Antigen synonyms:Natural killer cell lectin|CD159a antigen|MGC13374|Killer cell lectin like receptor subfamily C member 1|CD159 antigen-like family member A|KLRC1|CD159 antigen like family member A|CD159a|MGC59791
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:26 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human KLRC1 (amino acids 22 - 71)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to KLRC1 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: NKG2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human