Anti-OR2M3 Rabbit Polyclonal Antibody
Catalog # ANTIA100915-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Olfactory receptor 2M3
- Antigen symbol:OR2M3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q8NG83
- Antigen synonyms:OST003|Olfactory receptor family 2 subfamily M member 3|OR2M3P|OR2M3|OR2M3_HUMANOR2M6|Olfactory receptor 2M6|Olfactory receptor OR1-54
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:34 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human OR2M3 (amino acids 241 - 290)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to OR2M3 for IF and ELISA with samples derived from human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: OR2M3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human