Order Entry
Export
ContactUsLinkComponent
Anti-OR52E4 Rabbit Polyclonal Antibody
Anti-OR52E4 Rabbit Polyclonal Antibody
Catalog # ANTIA100383-100
Supplier:  Antibodies.com
New Product
undefined
Anti-OR52E4 Rabbit Polyclonal Antibody
Catalog # ANTIA100383-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    O52E4_HUMAN
  • Antigen symbol:
    OR52E4
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8NGH9
  • Antigen synonyms:
    Olfactory receptor OR11-55|Olfactory receptor, family 52, subfamily E, member 4|OR52E4|Olfactory receptor 52E4
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    35 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human OR52E4 (amino acids 190 - 239)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to OR52E4 for WB and ELISA with samples derived from human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000

Type: Primary
Antigen: OR52E4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human