Order Entry
Export
ContactUsLinkComponent
Anti-OR56A3 Rabbit Polyclonal Antibody
Anti-OR56A3 Rabbit Polyclonal Antibody
  ANTIA100371-100
New Product
undefined
Anti-OR56A3 Rabbit Polyclonal Antibody
  ANTIA100371-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    O56A3_HUMAN
  • Antigen symbol:
    OR56A3
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q8NH54
  • Antigen synonyms:
    Olfactory receptor family 56 subfamily A member 3 pseudogene|Olfactory receptor 56A6|OR56A2P|OR56A3P|Olfactory receptor 56A3|Olfactory receptor OR11-51|OR56A3|Olfactory receptor family 56 subfamily A member 2 pseudogene|Olfactory receptor, family 56, subfamily A, member 3
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    35 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human OR56A3 (amino acids 266 - 315)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Rabbit polyclonal antibody to OR56A3 for IF and ELISA with samples derived from human.

  • Validated Applications: IF, ELISA
  • Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:20000

Type: Primary
Antigen: OR56A3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human