Anti-Sodium bicarbonate transporter-like protein 11 Rabbit Polyclonal Antibody
Catalog # ANTIA100094-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Bicarbonate transporter related protein 1
- Antigen symbol:Sodium bicarbonate transporter-like protein 11
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q8NBS3
- Antigen synonyms:Bicarbonate transporter-related protein 1|Corneal endothelial dystrophy 2 autosomal recessive|FECD4|MGC126418|CHED2|MGC126419|dJ794I6.2|CDPD|BTR1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:99 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SLC4A11 (amino acids 276 - 325)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to SLC4A11 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Sodium bicarbonate transporter-like protein 11
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human