Anti-ICAM1 Mouse Monoclonal Antibody [clone: ABT-CD54]
Supplier: Antibodies.com
New Product
Certificates
About this item
Mouse monoclonal (ABT-CD54) antibody to CD54 for IHC with samples derived from human.
- Validated Applications: IHC
Type: Primary
Antigen: ICAM1
Clonality: Monoclonal
Clone: ABT-CD54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ANTIA100027-100
- ANTIA100027-50
- Antigen symbol:
- ICAM1
- ICAM1
- Antigen name:
- Antigen identified by monoclonal
- Antigen identified by monoclonal
- Antigen synonyms:
- Human rhinovirus receptor|CD54 antigen|Cell surface glycoprotein P3.58|BB2|CD54|CD_antigen=CD54|BB 2|CD 54|ICAM 1
- Human rhinovirus receptor|CD54 antigen|Cell surface glycoprotein P3.58|BB2|CD54|CD_antigen=CD54|BB 2|CD 54|ICAM 1
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- ABT-CD54
- ABT-CD54
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# P05362
- UniprotID# P05362
- Isotype:
- IgG
- IgG
- ImmunoChemistry:
- Yes
- Yes
- Immunogen:
- Synthetic peptide derived from human CD54
- Synthetic peptide derived from human CD54
- Form:
- Liquid
- Liquid
- Purification:
- Immunogen affinity purified
- Immunogen affinity purified
- Storage buffer:
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Supplied in Tris Buffered Saline with 0,5% Bovine Serum Albumin and 0,1% Kathon CG/ICP
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
- Tested applications:
- IHC
- IHC