Anti-LSP1 Rabbit Polyclonal Antibody
Catalog # ANTIA101390-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:47 kDa actin binding protein
- Antigen symbol:LSP1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P33241
- Antigen synonyms:Leufactin|LSP 1|52 kDa phosphoprotein|Leukocyte F actin binding protein|47 kDa actin-binding protein|LSP1|F actin binding and cytoskeleton associated protein|Leukocyte specific protein 1|Leufactin (leukocyte F-actin binding protein)
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:37 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human LSP1 (amino acids 104-153)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to LSP1 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: LSP1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human