Anti-MRGPRX4 Rabbit Polyclonal Antibody
ANTIA100976-100
New Product
- Antibody type:Primary
- Antigen name:G protein coupled receptor MRGX4
- Antigen symbol:MRGPRX4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# Q96LA9
- Antigen synonyms:Mas related G protein coupled receptor member X4|G protein coupled receptor SNSR5|MGC129754|G protein coupled receptor SNSR6|GPCR|Mas related G protein coupled-receptor family, member X4|MGC129753|Mas-related G-protein coupled receptor member X4|MAS related GPR, member X4
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:36 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human MRGX4 (amino acids 258-307)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to MRGX4 for WB and ELISA with samples derived from human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: MRGPRX4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human