Order Entry
Export
ContactUsLinkComponent
 
Anti-UGT2B7 Rabbit Polyclonal Antibody
  ANTIA8955-100
New Product
undefined
Anti-UGT2B7 Rabbit Polyclonal Antibody
  ANTIA8955-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    3, 3 4 catechol estrogen specific
  • Antigen symbol:
    UGT2B7
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P16662
  • Antigen synonyms:
    UDP glucuronosyltransferase 2B7|UDP glucuronosyltransferase 2 family, member B7|UD2B7_HUMAN|3,4 catechol estrogen specific UDPGT|UDP glucuronosyltransferase 2 family, polypeptide B7|UDP glucuronosyltransferase 2 family, member B9|4-catechol estrogen-specific UDPGT|UDP glucuronosyltransferase 2 family polypeptide B7
  • Storage buffer:
    Supplied in Phosphate buffered saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    70 kDa
  • Sequence:
    KVLVWAAEYSHWMNIKTILDELIQRGHEVTVLASSASILFDPNNSSALKIEIYPTSLTKTELENFIMQQIKRWSDLPKDTFWLYFSQVQEIMSIFGDITRKFCKDVVSNKKFMKKVQESRFDVIFADAIFPCSELLAELFNIPFVYSLSFSPGYTFEKHSGGFIFPPSYVPVVMSELTDQMTFMERVKNMIYVLYFDFWFEIFDMKKWDQFYSEVLGRPTTLSETMGKADVWLIRNSWNFQFPYPLLPNVDFVGGLHCKPAKPLPKEMEDFVQSSGENGVVVFSLGSMVSNMTEERANVIASALAQIPQKVLWRFDGNKPDTLGLNT
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at ‒20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 25-351 of human UGT2B7 (NP_001065.2)
  • Tested applications:
    ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

 

 

Rabbit polyclonal antibody to UGT2B7 for WB and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:2000, ICC/IF: 1:50 to 1:10

Type: Primary
Antigen: UGT2B7
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat