Anti-GAP43 Rabbit Polyclonal Antibody
Catalog # ANTIA94061-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Axonal membrane protein GAP 43
- Antigen symbol:GAP43
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P17677
- Antigen synonyms:Axonal membrane protein GAP-43|Growth-associated protein 43|Calmodulin binding protein P 57|Nerve growth related peptide GAP43|Growth Associated Protein 43|Nerve Growth Related Peptide|GAP43|B 50|F1
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:24 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human GAP43 around the phosphorylation site of Ser41 (amino acids 8-57)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to GAP43 (phospho Ser41) for IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: IF, ELISA
- Recommended dilutions: IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: GAP43
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat