Anti-alpha 2C Adrenergic Receptor Rabbit Polyclonal Antibody
ANTIA93945-100
New Product
- Antibody type:Primary
- Antigen name:Adra 2c
- Antigen symbol:alpha 2C Adrenergic Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P18825
- Antigen synonyms:ADRA2L2|Alpha 2C adrenergic receptor subtype C4|ADRA2RL2|ADRA2C|alpha 2C Adrenergic Receptor|Adrenoceptor alpha 2C|Adrenergic alpha 2C receptor|Alpha 2C adrenoceptor|ADRARL2
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:49 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Adrenergic Receptor alpha-2C (amino acids 336-385)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to Adrenergic Receptor alpha-2C for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: WB, ELISA
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:100, ELISA: 1:40000
Type: Primary
Antigen: alpha 2C Adrenergic Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat