Order Entry
Export
ContactUsLinkComponent
Anti-VE Cadherin Rabbit Polyclonal Antibody
Anti-VE Cadherin Rabbit Polyclonal Antibody
Catalog # ANTIA93936-100
Supplier:  Antibodies.com
New Product
undefined
Anti-VE Cadherin Rabbit Polyclonal Antibody
Catalog # ANTIA93936-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    7B 4
  • Antigen symbol:
    VE Cadherin
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P33151
  • Antigen synonyms:
    cadherin, vascular endothelial, 1|Cadherin 5 type 2|Cadherin 5, type 2, VE cadherin (vascular epithelium)|Cadherin 5, type 2 (vascular endothelium)|7B4|cadherin, vascular endothelial|Cadherin 5|7B4 antigen|CADH5_HUMAN
  • Storage buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
  • Molecular weight:
    87 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic phospho-peptide derived from human VE-Cadherin around the phosphorylation site of Tyr731
  • Tested applications:
    IHC
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rabbit polyclonal antibody to VE-Cadherin (phospho Tyr731) for WB, IHC and ELISA with samples derived from Human and Mouse.

  • Validated applications: WB, IHC, ELISA
  • Recommended dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:10000

Type: Primary
Antigen: VE Cadherin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse