Anti-alpha Adducin Rabbit Polyclonal Antibody
Catalog # ANTIA93909-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:ADD A
- Antigen symbol:alpha Adducin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P35611
- Antigen synonyms:ADDA|ADDA_HUMAN|Adducin 1 (alpha)|Erythrocyte adducin alpha subunit|ADD1|MGC3339|Erythrocyte adducin subunit alpha|alpha Adducin|Alpha-adducin
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:80 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ADD1 around the phosphorylation site of Thr445 (amino acids 411-460)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to ADD1 (phospho Thr445) for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: IHC, ELISA
- Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:1000
Type: Primary
Antigen: alpha Adducin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat