Anti-PKD1 (phospho Ser738) Rabbit Polyclonal Antibody
ANTIA93896-100
New Product
- Antibody type:Primary
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# Q15139, Q9BZL6, O94806
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:101 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic phospho-peptide derived from human PKD1 around the phosphorylation site of Ser738
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Pack type:Plastic vial
- Pk:100 µG
Rabbit polyclonal antibody to PKD1 (phospho Ser738) for IHC and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: IHC, ELISA
- Recommended dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat