Anti-p53 Rabbit Polyclonal Antibody
Catalog # ANTIA93560-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen NY-CO-13
- Antigen symbol:p53
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Form:Liquid
- Gene ID:UniprotID# P04637
- Antigen synonyms:P53_HUMAN|Tp53|BCC7|Cellular tumor antigen p53|Mutant tumor protein 53|LFS1|Phosphoprotein p53|FLJ92943|p53 tumor suppressor
- Storage buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7,4, with 150 mM NaCl, 0,02% Sodium Azide, and 50% Glycerol
- Molecular weight:43 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human p53 around the acetylated site of Lys379 (amino acids 344-393)
- Tested applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing acetylated peptide
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rabbit polyclonal antibody to p53 (acetyl Lys379) for IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated applications: IHC, IF, ELISA
- Recommended dilutions: IHC: 1:50 to 1:100, IF: 1:100 to 1:500, ELISA: 1:10000
Type: Primary
Antigen: p53
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat