Order Entry
Export
ContactUsLinkComponent
Anti-CD57 Mouse Monoclonal Antibody [Clone: TB01]
Anti-CD57 Mouse Monoclonal Antibody [Clone: TB01]
  ANTIA86858-100
New Product
undefined
Anti-CD57 Mouse Monoclonal Antibody [Clone: TB01]
  ANTIA86858-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    3-glucuronyltransferase
  • Antigen symbol:
    CD57
  • Clonality:
    Monoclonal
  • Clone:
    TB01
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9P2W7
  • Antigen synonyms:
    Beta 1 3 glucuronyltransferase 1|3-glucuronyltransferase 1|CD57|Beta-1|CD 57|B3gat1|CD57 antigen|Beta-1,3-glucuronyltransferase 1|B3GA1_HUMAN
  • Storage buffer:
    Supplied in Tris Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    A pool of neuroblastoma cell lines
  • Tested applications:
    IHC-P, IHC-Fr, ICC
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [TB01] antibody to CD57 for Flow Cytometry, WB, IHC-P, IHC-Fr and ICC with samples derived from Human.

  • Validated applications: Flow cytometry, WB, IHC-P, IHC-Fr, ICC
  • Recommended dilutions: Flow cytometry: 1 to 4 µg/ml, IHC: 5 to 10 µg/ml

Type: Primary
Antigen: CD57
Clonality: Monoclonal
Clone: TB01
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human