Anti-CD8 alpha Rat Monoclonal Antibody [Clone: 53-6.7]
Catalog # ANTIA86753-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:alpha polypeptide (p32)
- Antigen symbol:CD8 alpha
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:Liquid
- Gene ID:UniprotID# P01731
- Antigen synonyms:CD8 alpha|CD8A antigen|CD8A_HUMAN|CD8a|Leu2|CD8 antigen alpha polypeptide (p32)|CD8|CD8 antigen alpha polypeptide|CD8A molecule
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, without Sodium Azide (0,2 µm filter sterilised). Endotoxin level is less than 0,01 EU/µg of the protein, as determined by the LAL test
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Mouse spleen cells
- Tested applications:IHC-P, IHC-Fr, ICC, Functional studies
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Rat monoclonal [53-6.7] antibody to CD8 alpha for Flow Cytometry, IP, IHC-P, IHC-Fr, ICC and Functional Studies with samples derived from Mouse.
- Validated applications: Flow cytometry, IP, IHC-P, IHC-Fr, ICC, Functional Studies
- Recommended dilutions: Flow cytometry: 1.5 µg/ml, IHC-Fr: 1:1, 000
Type: Primary
Antigen: CD8 alpha
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse