Order Entry
Export
ContactUsLinkComponent
Anti-CD8 alpha Rat Monoclonal Antibody [Clone: 53-6.7] (PE (Phycoerythrin))
Anti-CD8 alpha Rat Monoclonal Antibody [Clone: 53-6.7] (PE (Phycoerythrin))
Catalog # ANTIA86752-100
Supplier:  ANTIBODIES.COM
New Product
undefined
Anti-CD8 alpha Rat Monoclonal Antibody [Clone: 53-6.7] (PE (Phycoerythrin))
Catalog # ANTIA86752-100
Supplier:  ANTIBODIES.COM
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    alpha polypeptide (p32)
  • Antigen symbol:
    CD8 alpha
  • Clonality:
    Monoclonal
  • Clone:
    53-6.7
  • Conjugation:
    PE (Phycoerythrin)
  • Flow cytometry:
    Yes
  • Host:
    Rat
  • Isotype:
    IgG2a kappa
  • Reactivity:
    Mouse
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P01731
  • Antigen synonyms:
    CD8 alpha|CD8A antigen|CD8A_HUMAN|CD8a|Leu2|CD8 antigen alpha polypeptide (p32)|CD8|CD8 antigen alpha polypeptide|CD8A molecule
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Concentration:
    0,5 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Mouse spleen cells
  • Purification:
    This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Rat monoclonal [53-6.7] antibody to CD8 alpha (PE) for Flow Cytometry with samples derived from Mouse.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: 2 µg/ml

Type: Primary
Antigen: CD8 alpha
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse