Anti-CD8 alpha Rat Monoclonal Antibody [Clone: 53-6.7] (APC (Allophycocyanin))
ANTIA86750-100
New Product
- Antibody type:Primary
- Antigen name:alpha polypeptide (p32)
- Antigen symbol:CD8 alpha
- Clonality:Monoclonal
- Clone:53-6.7
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Form:Liquid
- Gene ID:UniprotID# P01731
- Antigen synonyms:CD8 alpha|CD8A antigen|CD8A_HUMAN|CD8a|Leu2|CD8 antigen alpha polypeptide (p32)|CD8|CD8 antigen alpha polypeptide|CD8A molecule
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Concentration:0,5 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Mouse spleen cells
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Plastic vial
- Pk:100 µG
Rat monoclonal [53-6.7] antibody to CD8 alpha (APC) for Flow Cytometry with samples derived from Mouse.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 2 µg/ml
Type: Primary
Antigen: CD8 alpha
Clonality: Monoclonal
Clone: 53-6.7
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Mouse