Order Entry
Export
ContactUsLinkComponent
Anti-alpha Tubulin Mouse Monoclonal Antibody [Clone: TU-16]
Anti-alpha Tubulin Mouse Monoclonal Antibody [Clone: TU-16]
Catalog # ANTIA86709-100
Supplier:  Antibodies.com
New Product
undefined
Anti-alpha Tubulin Mouse Monoclonal Antibody [Clone: TU-16]
Catalog # ANTIA86709-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    alpha Tubulin
  • Antigen symbol:
    alpha Tubulin
  • Clonality:
    Monoclonal
  • Clone:
    TU-16
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgM
  • Reactivity:
    Pig,
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q71U36
  • Antigen synonyms:
    H2 ALPHA|Hum a tub2|ALS22|MGC171407|Hum a tub1|bA408E5.3|B ALPHA 1|Alpha-tubulin 1|LIS3
  • Storage buffer:
    Supplied in Tris Buffered Saline, pH 8,0, with 15 mM Sodium Azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Porcine brain microtubule protein MTP-1
  • Tested applications:
    IHC-P, ICC
  • Purification:
    Purified by precipitation and chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [TU-16] antibody to alpha Tubulin for IP, WB, IHC-P, ICC and ELISA with samples derived from Human, Mouse, Rat, Porcine and Many Others.

  • Validated applications: IP, WB, IHC-P, ICC, ELISA
  • Recommended dilutions: WB: 1 to 2 µg/ml, IHC-P: 10 µg/ml

Type: Primary
Antigen: alpha Tubulin
Clonality: Monoclonal
Clone: TU-16
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Human, Mouse, Rat, Porcine, Many Others