Anti-Cytokeratin 18 Mouse Monoclonal Antibody [Clone: C-04] (Biotinylated)
Catalog # ANTIA86659-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Cell proliferation inducing gene 46 protein
- Antigen symbol:Cytokeratin 18
- Clonality:Monoclonal
- Clone:C-04
- Conjugation:Biotin
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Hamster,Pig,Sheep,Goat,Human,Bovine,Dog,Rat
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05783
- Antigen synonyms:CK18|Cell proliferation inducing protein 46|Cytokeratin 18|CYK18|Cytokeratin endo B|Cell proliferation-inducing gene 46 protein|CK 18|CK-18|CYK 18
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
- Storage temperature:Store at 2...8 °C. Do not freeze!
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cytoskeleton preparation of epidermal carcinoma cell line A431
- Tested applications:IHC-P, ICC
- Purification:This antibody is conjugated with Biotin-LC-NHS under optimum conditions. The reagent is free of unconjugated Biotin.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [C-04] antibody to Cytokeratin 18 (Biotin) for IHC-P, Flow Cytometry, IP, WB, ICC and ELISA with samples derived from Human, Rat, Sheep, Goat, Hamster, Bovine, Canine and Porcine.
- Validated applications: IHC-P, Flow cytometry, IP, WB, ICC, ELISA
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml, WB: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 18
Clonality: Monoclonal
Clone: C-04
Conjugation: Biotin
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Sheep, Goat, Hamster, Bovine, Dog, Porcine