Anti-HLA B7 Mouse Monoclonal Antibody [Clone: BB7.1] (PE (Phycoerythrin))
Catalog # ANTIA86655-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Abacavir hypersensitivity, susceptibility to
- Antigen symbol:HLA B7
- Clonality:Monoclonal
- Clone:BB7.1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Form:Liquid
- Antigen synonyms:HLA class I histocompatibility antigen B 7 alpha chain|HLA class I histocompatibility antigen, B-15 alpha chain|HLA class I histocompatibility antigen, B-18 alpha chain|HLA B|HLA class I histocompatibility antigen, B-13 alpha chain|HLA class I histocompatibility antigen, B-27 alpha chain|Drug-induced liver injury due to flucloxacillin|HLA B7|HLA class I histocompatibility antigen, B-14 alpha chain
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Papain solubilised HLA-A2, B7
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [BB7.1] antibody to HLA B7 (PE) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: HLA B7
Clonality: Monoclonal
Clone: BB7.1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates