Anti-CD3 zeta (phospho Tyr142) Mouse Monoclonal Antibody [Clone: EM-54]
Catalog # ANTIA86525-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:4930549J05Rik
- Antigen symbol:CD3 zeta
- Clonality:Monoclonal
- Clone:EM-54
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P20963
- Antigen synonyms:CD3 antigen, zeta polypeptide, isoform CRA_b|CD247|CD3 antigen, zeta subunit|CD247 antigen|Cd3|A430104F18Rik|AW552088|CD3 zeta|Cd247 molecule
- Modification:phospho Tyr142
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:CP1 (pTyr142) - KLH
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [EM-54] antibody to CD3 zeta (phospho Tyr142) for Flow Cytometry and WB with samples derived from Human and Mouse.
- Validated applications: Flow cytometry, WB
- Recommended dilutions: Flow cytometry: 1 to 4 µg/ml
Type: Primary
Antigen: CD3 zeta
Clonality: Monoclonal
Clone: EM-54
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse