Anti-pan Cytokeratin Mouse Monoclonal Antibody [Clone: C-11] (APC (Allophycocyanin))
Catalog # ANTIA86441-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:pan Cytokeratin
- Antigen symbol:pan Cytokeratin
- Clonality:Monoclonal
- Clone:C-11
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Goat,Human,Rat,Mouse
- Form:Liquid
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Concentration:0,1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Keratin-enriched preparation from human epidermoid carcinoma cell line A431
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [C-11] antibody to pan Cytokeratin (APC) for Flow Cytometry with samples derived from Human, Rat, Goat and Mouse.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 µg/ml
Type: Primary
Antigen: pan Cytokeratin
Clonality: Monoclonal
Clone: C-11
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Rat, Goat, Mouse