Anti-Cytokeratin 7 + 17 Mouse Monoclonal Antibody [Clone: C-46]
Catalog # ANTIA86418-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:Cytokeratin 7 + 17
- Clonality:Monoclonal
- Clone:C-46
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Bovine
- Western blot:Yes
- Form:Liquid
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cytoskeleton preparation of HeLa human cervix carcinoma cell line
- Tested applications:IHC-P, ICC
- Purification:Protein A chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [C-46] antibody to Cytokeratin 7 + 17 for IP, WB, IHC-P and ICC with samples derived from Human, Porcine and Bovine.
- Validated applications: IP, WB, IHC-P, ICC
- Recommended dilutions: IHC-P: 10 µg/ml, WB: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 7 + 17
Clonality: Monoclonal
Clone: C-46
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Porcine, Bovine