Anti-OPAL1 Mouse Monoclonal Antibody [Clone: OPAL1-01] (PE (Phycoerythrin))
ANTIA86383-100
New Product
- Antibody type:Primary
- Antigen name:C10orf26
- Antigen symbol:OPAL1
- Clonality:Monoclonal
- Clone:OPAL1-01
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q9NX94
- Antigen synonyms:OPA1L|OPA1L_HUMAN|FLJ20154|Outcome predictor in acute leukemia 1|OPAL1|WW domain binding protein 1-like|WBP1L
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Concentration:0,1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment of human OPAL1 (amino acids 152-342)
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [OPAL1-01] antibody to OPAL1 (PE) for Flow Cytometry with samples derived from Human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 5 µg/ml
Type: Primary
Antigen: OPAL1
Clonality: Monoclonal
Clone: OPAL1-01
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human