Anti-CD94 Mouse Monoclonal Antibody [Clone: HP-3D9] (APC (Allophycocyanin))
Catalog # ANTIA86321-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:CD 94
- Antigen symbol:CD94
- Clonality:Monoclonal
- Clone:HP-3D9
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q13241
- Antigen synonyms:CD94 antigen|KLRD 1|KLRD1|KLRD1_HUMAN|KLRD1 protein|Killer cell lectin-like receptor subfamily D member 1|KP 43|Killer cell lectin like receptor subfamily D member 1|CD94
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cultured human NK cells
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [HP-3D9] antibody to CD94 (APC) for Flow Cytometry with samples derived from Human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD94
Clonality: Monoclonal
Clone: HP-3D9
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human