Anti-DR5 Mouse Monoclonal Antibody [Clone: DR5-01-1] (PE (Phycoerythrin))
Catalog # ANTIA86274-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Apoptosis inducing protein TRICK2A/2B
- Antigen symbol:DR5
- Clonality:Monoclonal
- Clone:DR5-01-1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# O14763
- Antigen synonyms:CD262 antigen|Fas like protein|Death domain containing receptor for TRAIL/Apo 2L|CD262|KILLER|Death receptor 5|Cytotoxic TRAIL receptor 2|Apoptosis inducing receptor TRAIL R2|DR5
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Concentration:0,1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fusion protein of human IgG heavy chain and extracellular domain of DR5
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [DR5-01-1] antibody to DR5 (PE) for Flow Cytometry with samples derived from Human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: 1 to 5 µg/ml
Type: Primary
Antigen: DR5
Clonality: Monoclonal
Clone: DR5-01-1
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human