Anti-CD163 Mouse Monoclonal Antibody [Clone: GHI/61] (PE (Phycoerythrin))
Catalog # ANTIA86217-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:C163A_HUMAN
- Antigen symbol:CD163
- Clonality:Monoclonal
- Clone:GHI/61
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# Q86VB7
- Antigen synonyms:CD163 antigen|CD163 molecule|CD163|M130 antigen precursor|Hemoglobin scavenger receptor|M130|MM130|CD 163|Macrophage associated antigen
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Hairy cell leukemia cells
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [GHI/61] antibody to CD163 (PE) for Flow Cytometry with samples derived from Human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD163
Clonality: Monoclonal
Clone: GHI/61
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human