Anti-CD36 Mouse Monoclonal Antibody [Clone: CB38 (NL07)] (PE (Phycoerythrin))
Catalog # ANTIA86175-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:Adipocyte membrane protein
- Antigen symbol:CD36
- Clonality:Monoclonal
- Clone:CB38 (NL07)
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgM kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P16671
- Antigen synonyms:CHDS7|CD36|BDPLT10|CD36 molecule (thrombospondin receptor)|CD36 molecule|Cluster determinant 36|CD36 antigen|CD36_HUMAN|CD36 antigen (collagen type I receptor, thrombospondin receptor)
- Storage buffer:Supplied in Tris Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Living human myeloid cells
- Purification:This antibody is conjugated with PE under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [CB38 (NL07)] antibody to CD36 (PE) for Flow Cytometry with samples derived from Human.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD36
Clonality: Monoclonal
Clone: CB38 (NL07)
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human