Order Entry
Export
ContactUsLinkComponent
 
Anti-CD5 Mouse Monoclonal Antibody [Clone: CRIS1]
  ANTIA86153-100
New Product
undefined
Anti-CD5 Mouse Monoclonal Antibody [Clone: CRIS1]
  ANTIA86153-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    CD 5
  • Antigen symbol:
    CD5
  • Clonality:
    Monoclonal
  • Clone:
    CRIS1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2a
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P06127
  • Antigen synonyms:
    CD5|CD5 molecule|CD5 antigen (p56 62)|CD5 antigen|LEU1|OTTHUMP00000236973|CD5_HUMAN|Lymphocyte antigen T1/Leu-1|LyA
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,4, with 15 mM Sodium Azide
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Stimulated human leukocytes
  • Tested applications:
    IHC-Fr
  • Purification:
    Protein A chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [CRIS1] antibody to CD5 for Flow Cytometry, IP, WB, IHC-Fr and ELISA with samples derived from Human.

  • Validated applications: Flow cytometry, IP, WB, IHC-Fr, ELISA
  • Recommended dilutions: Flow cytometry: 1 µg/ml, WB: 1 to 2 µg/ml

Type: Primary
Antigen: CD5
Clonality: Monoclonal
Clone: CRIS1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human