Anti-CD48 Mouse Monoclonal Antibody [Clone: MEM-102] (APC (Allophycocyanin))
Catalog # ANTIA86025-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:Antigen CD48
- Antigen symbol:CD48
- Clonality:Monoclonal
- Clone:MEM-102
- Conjugation:APC (Allophycocyanin)
- Flow cytometry:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Non-human primates,Human
- Form:Liquid
- Gene ID:UniprotID# P09326
- Antigen synonyms:BCM1 surface antigen|B cell membrane protein|BLAST|BLAST1|BCM 1 surface antigen|BCM1|B lymphocyte activation marker BLAST 1|B-lymphocyte activation marker BLAST-1|B-cell activation marker
- Storage buffer:Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
- Storage temperature:Store in the dark at 2...8 °C. Do not freeze!
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Raji human Burkitt's lymphoma cell line
- Purification:This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
- Pack type:Glass bottle
- Pk:100 T
Specifications
About this item
Mouse monoclonal [MEM-102] antibody to CD48 (APC) for Flow Cytometry with samples derived from Human and Non-Human Primates.
- Validated applications: Flow cytometry
- Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension
Type: Primary
Antigen: CD48
Clonality: Monoclonal
Clone: MEM-102
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Non-Human Primates