Order Entry
Export
ContactUsLinkComponent
Anti-CD45RA Mouse Monoclonal Antibody [Clone: MEM-56] (APC (Allophycocyanin))
Anti-CD45RA Mouse Monoclonal Antibody [Clone: MEM-56] (APC (Allophycocyanin))
Catalog # ANTIA86002-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD45RA Mouse Monoclonal Antibody [Clone: MEM-56] (APC (Allophycocyanin))
Catalog # ANTIA86002-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    B220
  • Antigen symbol:
    CD45RA
  • Clonality:
    Monoclonal
  • Clone:
    MEM-56
  • Conjugation:
    APC (Allophycocyanin)
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Form:
    Liquid
  • Antigen synonyms:
    CD45R|CD45|CD45RA|GP180|Leukocyte common antigen|L-CA|LCA|CD45 antigen|EC 3.1.3.48
  • Storage buffer:
    Supplied in Phosphate Buffered Saline with 15 mM Sodium Azide
  • Storage temperature:
    Store in the dark at 2...8 °C. Do not freeze!
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human thymocytes and T lymphocytes
  • Purification:
    This antibody is conjugated with APC under optimum conditions. The conjugate is purified by size-exclusion chromatography.
  • Pack type:
    Glass bottle
  • Pk:
    100 T

Specifications

About this item

Mouse monoclonal [MEM-56] antibody to CD45RA (APC) for Flow Cytometry with samples derived from Human.

  • Validated applications: Flow cytometry
  • Recommended dilutions: Flow cytometry: This reagent is designed for analysis of human blood cells using 10 µl reagent/100 µl of whole blood or 106 cells in a suspension

Type: Primary
Antigen: CD45RA
Clonality: Monoclonal
Clone: MEM-56
Conjugation: APC (Allophycocyanin)
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human