Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 20 Rabbit Polyclonal Antibody
Anti-Cytokeratin 20 Rabbit Polyclonal Antibody
Catalog # ANTIA92935-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Cytokeratin 20 Rabbit Polyclonal Antibody
Catalog # ANTIA92935-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD20
  • Antigen symbol:
    Cytokeratin 20
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P35900
  • Antigen synonyms:
    Cytokeratin-20|Keratin|CK-20|CK20|CK 20|Cytokeratin20|KA20|K1C20_HUMAN|Cytokeratin 20
  • Storage buffer:
    Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
  • Molecular weight:
    48 kDa
  • Sequence:
    KYEVMAQKNLQEAKEQFERQTAVLQQQVTVNTEELKGTEVQLTELRRTSQSLEIELQSHLSMKESLEHTLEETKARYSSQLANLQSLLSSLEAQLMQIRSNMERQNNEYHILLDIKTRLEQEIATYRRLLEGEDVKTTEYQLSTLEERDIKKTRKIKTVVQEVVDGKVVSSEVKEVEENI
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fusion protein containing a sequence corresponding to amino acids 245-424 of human Cytokeratin 20 (Cytokeratin 20 (KRT20)) (NP_061883.1)
  • Tested applications:
    IHC, ICC/IF
  • Purification:
    Affinity purification
  • Pack type:
    Plastic vial
  • Pk:
    100 µl

Specifications

About this item

Rabbit polyclonal antibody to Cytokeratin 20 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.

  • Validated applications: WB, IHC, ICC/IF
  • Recommended dilutions: WB: 1:500 to 1:2000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200

Type: Primary
Antigen: Cytokeratin 20
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat