Anti-IRE1 Rabbit Polyclonal Antibody
ANTIA92812-100
New Product
- Antibody type:Primary
- Antigen name:Endoplasmic reticulum (ER) to nucleus signalling 1
- Antigen symbol:IRE1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# O75460
- Antigen synonyms:ERN1_HUMAN|Endoribonuclease|Endoplasmic reticulum-to-nucleus signaling 1|hIRE1p|ERN 1|Endoplasmic reticulum to nucleus signaling 1|hIRE 1p|ER to nucleus signaling 1|Ern1
- Storage buffer:Supplied in Phosphate Buffered Saline, pH 7,3, with 50% Glycerol and 0,01% Thiomersal
- Molecular weight:130 kDa
- Sequence:RHSFS
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A synthetic phosphorylated peptide around S724 of human IRE1 (NP_001424.3)
- Tested applications:IHC, ICC/IF
- Purification:Affinity purification
- Pack type:Plastic vial
- Pk:100 µl
Rabbit polyclonal antibody to IRE1 for WB, IHC and ICC/IF with samples derived from Human, Mouse and Rat.
- Validated applications: WB, IHC, ICC/IF
- Recommended dilutions: WB: 1:500 to 1:1000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200
Type: Primary
Antigen: IRE1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat