Order Entry
Export
ContactUsLinkComponent
Anti-Cytokeratin 20 Rabbit Recombinant Antibody [clone: KRT20/4380R]
Anti-Cytokeratin 20 Rabbit Recombinant Antibody [clone: KRT20/4380R]
Catalog # ANTIA278633-100
Supplier:  Antibodies.com
New Product
undefined
Anti-Cytokeratin 20 Rabbit Recombinant Antibody [clone: KRT20/4380R]
Catalog # ANTIA278633-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    CD20
  • Antigen symbol:
    Cytokeratin 20
  • Clonality:
    Recombinant
  • Clone:
    KRT20/4380R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P35900
  • Antigen synonyms:
    Cytokeratin-20|Keratin|CK-20|CK20|CK 20|Cytokeratin20|KA20|K1C20_HUMAN|Cytokeratin 20
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    46 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 324-424, of human Cytokeratin 20. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [KRT20/4380R] antibody to Cytokeratin 20 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Cytokeratin 20
Clonality: Recombinant
Clone: KRT20/4380R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human