Anti-Arginase 1 Rabbit Recombinant Antibody [clone: ARG1/6790R]
Catalog # ANTIA278600-100
Supplier: Antibodies.com
New Product
Specifications
- Antibody type:Primary
- Antigen name:A I
- Antigen symbol:Arginase 1
- Clonality:Recombinant
- Clone:ARG1/6790R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05089
- Antigen synonyms:Al|ARG 1|Arginase-1|Arginase 1|Arginase type I|ARGI1_HUMAN|Arginase, liver|Arginase liver|arg1
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:35 - 38 kDa
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment, around amino acids 300-400, of human Arginase 1. The exact sequence is proprietary.
- Tested applications:IHC
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Recombinant rabbit monoclonal [ARG1/6790R] antibody to Arginase 1 for WB and IHC with samples derived from human.
- Validated Applications: WB, IHC
- Recommended Dilutions: WB: 2 to 4 µg/ml, IHC-P: 2 to 4 µg/ml
Type: Primary
Antigen: Arginase 1
Clonality: Recombinant
Clone: ARG1/6790R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human