Anti-EGFR Rabbit Recombinant Antibody [clone: EGFR/6390R]
ANTIA277974-100
New Product
- Antibody type:Primary
- Antigen name:9030024J15Rik
- Antigen symbol:EGFR
- Clonality:Recombinant
- Clone:EGFR/6390R
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P00533
- Antigen synonyms:EGFR_HUMAN|EGF R|Cell growth inhibiting protein 40 |EGFR|Cell proliferation inducing protein 61 |Avian erythroblastic leukemia viral (v erb b) oncogene homolog |AI552599|Epidermal growth factor receptor|Avian erythroblastic leukemia viral (verbb) oncogene homolog
- Storage buffer:Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
- Molecular weight:~170 kDa (wild type)/~145 kDa (vIII variant)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:200 µg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant fragment corresponding to the extracellular domain, around amino acids 25-645, of human EGFR protein. The exact sequence is proprietary.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Recombinant rabbit monoclonal [EGFR/6390R] antibody to EGFR for IHC-P with samples derived from human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: EGFR
Clonality: Recombinant
Clone: EGFR/6390R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human