Order Entry
Export
ContactUsLinkComponent
Anti-HOXB2 Mouse Monoclonal Antibody [clone: PCRP-HOXB2-1F2]
Anti-HOXB2 Mouse Monoclonal Antibody [clone: PCRP-HOXB2-1F2]
Catalog # ANTIA277644-100
Supplier:  Antibodies.com
New Product
undefined
Anti-HOXB2 Mouse Monoclonal Antibody [clone: PCRP-HOXB2-1F2]
Catalog # ANTIA277644-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    homeo box 2H
  • Antigen symbol:
    HOXB2
  • Clonality:
    Monoclonal
  • Clone:
    PCRP-HOXB2-1F2
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P14652
  • Antigen synonyms:
    Homeobox protein Hox-2H|Hox 2 8|HOX2H|Homeobox protein Hox-2.8|Homeobox protein Hox-B2|HOX2|homeo box B2|homeobox B2|Hox 2H
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline with 0,05% BSA and 0,05% sodium azide
  • Molecular weight:
    37,9 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    200 µg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant fragment, around amino acids 114-220, of human HOXB2 protein. The exact sequence is proprietary.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [PCRP-HOXB2-1F2] antibody to HOXB2 for ELISA, IP, Flow Cytometry, IF, WB and IHC-P with samples derived from human.

  • Validated Applications: ELISA, IP, Flow Cytometry, IF, WB, IHC-P
  • Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: HOXB2
Clonality: Monoclonal
Clone: PCRP-HOXB2-1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human