Anti-CD59 Mouse Monoclonal Antibody [clone: 193-27]
Catalog # ANTIA254356-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen name:16.3A5
- Antigen symbol:CD59
- Clonality:Monoclonal
- Clone:193-27
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgM kappa
- Reactivity:Human
- Form:Liquid
- Gene ID:UniprotID# P13987
- Antigen synonyms:CD59 antigen|1F5|CD59 antigen complement regulatory protein |CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344)|CD 59|CD59|20 kDa homologous restriction factor|CD59 antigen p18 20|1F5 antigen
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Stimulated human leukocytes.
- Tested applications:Functional studies
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [193-27] antibody to CD59 for Functional Studies, Flow Cytometry and IF with samples derived from human.
- Validated Applications: Functional Studies, Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: 193-27
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgM kappa
Reactivity: Human