Anti-Cytokeratin 8 + 18 Mouse Monoclonal Antibody [clone: C-51]
Catalog # ANTIA254075-100
Supplier: ANTIBODIES.COM
New Product
Specifications
- Antibody type:Primary
- Antigen symbol:Cytokeratin 8 + 18
- Clonality:Monoclonal
- Clone:C-51
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Pig,Monkey,Sheep,Human,Bovine
- Western blot:Yes
- Form:Liquid
- Gene ID:UniprotID# P05787, P05783
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Molecular weight:52,5 kDa (CK8)/45 kDa (CK18)
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:Cytoskeleton preparation from HeLa cells.
- Tested applications:IHC-P
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Specifications
About this item
Mouse monoclonal [C-51] antibody to Cytokeratin 8 + 18 for Flow Cytometry, IF, WB and IHC-P with samples derived from human,monkey, bovine, sheep and porcine.
- Validated Applications: Flow Cytometry, IF, WB, IHC-P
- Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml, WB: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Cytokeratin 8 + 18
Clonality: Monoclonal
Clone: C-51
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human;Monkey;Bovine;Sheep;Pig