Anti-Enterobacter aerogenes Mouse Monoclonal Antibody [clone: 57/82]
ANTIA254035-100
New Product
- Antibody type:Primary
- Antigen symbol:Enterobacter aerogenes
- Clonality:Monoclonal
- Clone:57/82
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG kappa
- Reactivity:Enterobacter aerogenes
- Form:Liquid
- Storage buffer:Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
- Storage temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Concentration:1 mg/ml
- Shipping temperature:Shipped on blue ice at 4 °C
- Immunogen:A crude sonicate of Enterobacter aerogenes.
- Purification:Protein A/G chromatography
- Pack type:Plastic vial
- Pk:100 µG
Mouse monoclonal [57/82] antibody to Enterobacter aerogenes for ELISA and IF with samples derived from Enterobacter aerogenes.
- Validated Applications: ELISA, IF
- Recommended Dilutions: ELISA: 1 to 5 µg/ml, IF: 1 to 2 µg/ml
Type: Primary
Antigen: Enterobacter aerogenes
Clonality: Monoclonal
Clone: 57/82
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG kappa
Reactivity: Enterobacter aerogenes