Order Entry
Export
ContactUsLinkComponent
Anti-CD59 Mouse Monoclonal Antibody [clone: BRA-10G]
Anti-CD59 Mouse Monoclonal Antibody [clone: BRA-10G]
Catalog # ANTIA253919-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD59 Mouse Monoclonal Antibody [clone: BRA-10G]
Catalog # ANTIA253919-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    16.3A5
  • Antigen symbol:
    CD59
  • Clonality:
    Monoclonal
  • Clone:
    BRA-10G
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P13987
  • Antigen synonyms:
    CD59 antigen|1F5|CD59 antigen complement regulatory protein |CD59 antigen p18-20 (antigen identified by monoclonal antibodies 16.3A5, EJ16, EJ30, EL32 and G344)|CD 59|CD59|20 kDa homologous restriction factor|CD59 antigen p18 20|1F5 antigen
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    20 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Human K562 tumor cells.
  • Tested applications:
    Functional studies
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Mouse monoclonal [BRA-10G] antibody to CD59 for Functional Studies, Flow Cytometry and IF with samples derived from human.

  • Validated Applications: Functional Studies, Flow Cytometry, IF
  • Recommended Dilutions: Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: CD59
Clonality: Monoclonal
Clone: BRA-10G
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human