Order Entry
Export
ContactUsLinkComponent
Anti-CD47 Mouse Monoclonal Antibody [clone: IAP/964]
Anti-CD47 Mouse Monoclonal Antibody [clone: IAP/964]
  ANTIA253909-100
New Product
undefined
Anti-CD47 Mouse Monoclonal Antibody [clone: IAP/964]
  ANTIA253909-100
New Product

 

  • Antibody type:
    Primary
  • Antigen name:
    Antigen identified by monoclonal
  • Antigen symbol:
    CD47
  • Clonality:
    Monoclonal
  • Clone:
    IAP/964
  • Conjugation:
    Unconjugated
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q08722
  • Antigen synonyms:
    IAP|CD47_HUMAN|CD47|CD 47|CD47 antigen|CD47 molecule|Antigenic surface determinant protein OA3|CD47 glycoprotein|CD47 antigen (Rh-related antigen, integrin-associated signal transducer)
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    ~50 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human IAP protein.
  • Tested applications:
    Functional studies
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

 

 

Mouse monoclonal [IAP/964] antibody to CD47 for Functional Studies, WB, Flow Cytometry and IF with samples derived from human.

  • Validated Applications: Functional Studies, WB, Flow Cytometry, IF
  • Recommended Dilutions: WB: 1 to 2 µg/ml, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: CD47
Clonality: Monoclonal
Clone: IAP/964
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human