Order Entry
Export
ContactUsLinkComponent
Anti-CD44 Rabbit Recombinant Antibody [clone: HCAM/6459R]
Anti-CD44 Rabbit Recombinant Antibody [clone: HCAM/6459R]
Catalog # ANTIA253905-100
Supplier:  Antibodies.com
New Product
undefined
Anti-CD44 Rabbit Recombinant Antibody [clone: HCAM/6459R]
Catalog # ANTIA253905-100
Supplier:  Antibodies.com
New Product

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    BA-1
  • Antigen symbol:
    CD44
  • Clonality:
    Recombinant
  • Clone:
    HCAM/6459R
  • Conjugation:
    Unconjugated
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG kappa
  • Reactivity:
    Human
  • Form:
    Liquid
  • Gene ID:
    UniprotID# P16070
  • Antigen synonyms:
    CD44_HUMAN|CD44|CDw44|CD44 molecule (Indian blood group)|chondroitin sulfate proteoglycan 8|CD 44|Cell surface glycoprotein CD44|CD44 antigen|CD44 molecule
  • Storage buffer:
    Supplied in 10 mM phosphate buffered saline; without sodium azide and carrier free
  • Molecular weight:
    80 - 95 kDa
  • Storage temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Concentration:
    1 mg/ml
  • Shipping temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Recombinant full-length human CD44 protein.
  • Tested applications:
    IHC-P
  • Purification:
    Protein A/G chromatography
  • Pack type:
    Plastic vial
  • Pk:
    100 µG

Specifications

About this item

Recombinant rabbit monoclonal [HCAM/6459R] antibody to CD44 for IHC-P with samples derived from human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: CD44
Clonality: Recombinant
Clone: HCAM/6459R
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG kappa
Reactivity: Human