Anti-CD38 Rabbit Recombinant Antibody [clone: DM28] (Biotin)
Supplier: Antibodies.com
New Product
Certificates
About this item
Recombinant rabbit monoclonal [DM28] antibody to CD38 (Biotin) for ELISA and Flow Cytometry with samples derived from human.
- Validated Applications: ELISA, Flow Cytometry
- Recommended Dilutions: ELISA: 1:5,000 to 10,000, Flow Cytometry: 1:100
Type: Primary
Antigen: CD38
Clonality: Recombinant
Clone: DM28
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Specifications
- Catalog No:
- ANTIA317428-500
- ANTIA317428-100
- Antigen symbol:
- CD38
- CD38
- Antigen name:
- Acute lymphoblastic leukemia cells antigen CD38
- Acute lymphoblastic leukemia cells antigen CD38
- Antigen synonyms:
- ADPRC 1|ADP-ribosyl cyclase 1|CD38 antigen|ADPRC1|cADPr hydrolase 1|ADP ribosyl cyclase/cyclic ADP-ribose hydrolase|ADP ribosyl cyclase|ADP ribosyl cyclase 1|CD 38
- ADPRC 1|ADP-ribosyl cyclase 1|CD38 antigen|ADPRC1|cADPr hydrolase 1|ADP ribosyl cyclase/cyclic ADP-ribose hydrolase|ADP ribosyl cyclase|ADP ribosyl cyclase 1|CD 38
- Antibody type:
- Primary
- Primary
- Clonality:
- Recombinant
- Recombinant
- Conjugation:
- Biotin
- Biotin
- Clone:
- DM28
- DM28
- Reactivity:
- Human
- Human
- Host:
- Rabbit
- Rabbit
- Gene ID:
- UniprotID# P28907
- UniprotID# P28907
- Isotype:
- IgG
- IgG
- ELISA:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Form:
- Lyophilised
- Lyophilised
- Purification:
- Affinity chromatography
- Affinity chromatography
- Concentration:
- Reconstitution dependent
- Reconstitution dependent
- Storage buffer:
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
- Lyophilised from sterile phosphate buffered saline, pH 7,4. Normally 5 - 8% Trehalose is added as a protectant before lyophilisation
- Storage temperature:
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipped at 4 °C. Lyophilised: Store at –20...–80 °C. Reconstituted: Aliquot and store at –80 °C. Product is stable for one year. Avoid freeze/thaw cycles.
- Shipping temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C